Di-tert-butyl L-aspartate hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: L-Aspartic Acid Di-tert-butyl Ester HydrochlorideH-Asp(OtBu)-OtBu.HCl

D4422-25G D4422-5G
TCD4422-5GEA 62.52 USD
TCD4422-5G TCD4422-25G
Di-tert-butyl L-aspartate hydrochloride ≥98.0% (by HPLC, titration analysis)
Di-tert-butyl L-aspartate hydrochloride

CAS Number: 1791-13-5
MDL Number: MFCD00034851
Molecular Formula: C12H23NO4
Molecular Weight: 281.78
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 150
Specific rotation [a]20/D: 7 deg (C=1, MeOH)
Storage Temperature: 0-10°C


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: L-Aspartic Acid Di-tert-butyl Ester Hydrochloride, H-Asp(OtBu)-OtBu.HCl

Formula: C₁₂H₂₃NO₄
MW: 281.78 g/mol
Melting Pt: 150 °C
MDL Number: MFCD00034851
CAS Number: 1791-13-5
https://us.vwr.com/stibo/web/std.lang.all/65/31/28986531.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR