Dimethyl decahydro-1,4-naphthalenedicarboxylate (mixture of isomers) ≥97.0% (by GC)
Supplier: TCI America
Synonyms:
Decahydro-1,4-naphthalenedicarboxylic Acid Dimethyl Ester
D4399-1G
TCD4399-1GEA
399
USD
TCD4399-1G
Dimethyl decahydro-1,4-naphthalenedicarboxylate (mixture of isomers) ≥97.0% (by GC)
Dimethyl decahydro-1,4-naphthalenedicarboxylate (mixture of isomers)
CAS Number: 3068-03-9
Molecular Formula: C14H22O4
Molecular Weight: 254.33
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 178
Specific Gravity (20/20): 1.13
Other Product Names / Synonyms: Decahydro-1,4-naphthalenedicarboxylic Acid Dimethyl Ester
Formula:
C₁₄H₂₂O₄ MW: 254.33 g/mol Boiling Pt: 178 °C Density: 1.13 (20 °C) |
CAS Number:
3068-03-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...