Dimethyl [2-oxo-3-[3-(trifluoromethyl)phenoxy]propyl]phosphonate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: [2-Oxo-3-[3-(trifluoromethyl)phenoxy]propyl]phosphonic Acid Dimethyl Ester

D4397-1G
TCD4397-1GEA 451 USD
TCD4397-1G
Dimethyl [2-oxo-3-[3-(trifluoromethyl)phenoxy]propyl]phosphonate ≥98.0% (by GC)
Dimethyl [2-oxo-3-[3-(trifluoromethyl)phenoxy]propyl]phosphonate

CAS Number: 54094-19-8
MDL Number: MFCD09054713
Molecular Formula: C12H14F3O5P
Molecular Weight: 326.21
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.36


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: [2-Oxo-3-[3-(trifluoromethyl)phenoxy]propyl]phosphonic Acid Dimethyl Ester

Formula: C₁₂H₁₄F₃O₅P
MW: 326.21 g/mol
Density: 1.36 (20 °C)
MDL Number: MFCD09054713
CAS Number: 54094-19-8
https://us.vwr.com/stibo/web/std.lang.all/64/73/28986473.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR