Decahydro-1,4-naphthalenedicarboxylic acid (mixture of isomers) ≥96.0% (by GC, titration analysis)
Supplier: TCI America
D4383-1G
TCD4383-1GEA
348
USD
TCD4383-1G
Decahydro-1,4-naphthalenedicarboxylic acid (mixture of isomers) ≥96.0% (by GC, titration analysis)
Decahydro-1,4-naphthalenedicarboxylic acid (mixture of isomers)
CAS Number: 879360-14-2
Molecular Formula: C12H18O4
Molecular Weight: 226.27
Purity/Analysis Method:
Form: Crystal
Formula:
C₁₂H₁₈O₄ |
CAS Number:
879360-14-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...