1,3-Dimethoxy-4-nitrobenzene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2,4-Dimethoxy-1-nitrobenzene2,4-Dimethoxynitrobenzene

D4382-5G
TCD4382-5GEA 83 USD
TCD4382-5G
1,3-Dimethoxy-4-nitrobenzene ≥98.0% (by GC)
1,3-Dimethoxy-4-nitrobenzene

CAS Number: 4920-84-7
MDL Number: MFCD00024210
Molecular Formula: C8H9NO4
Molecular Weight: 183.16
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 76

Formula: C₈H₉NO₄
MW: 183.16 g/mol
Melting Pt: 76 °C
MDL Number: MFCD00024210
CAS Number: 4920-84-7
https://us.vwr.com/stibo/web/std.lang.all/64/43/28986443.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR