Diethyl-2-bromo-2-methylmalonate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Diethyl 2-bromo-2-methylmalonate2-Bromo-2-methylmalonic Acid Diethyl Ester

D4368-25G
TCD4368-25GEA 112.67 USD
TCD4368-25G
Diethyl-2-bromo-2-methylmalonate ≥98.0% (by GC, titration analysis)
Diethyl-2-bromo-2-methylmalonate

CAS Number: 29263-94-3
MDL Number: MFCD00009125
Molecular Formula: C8H13BrO4
Molecular Weight: 253.09
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 59
Flash Point (°C): 113
Specific Gravity (20/20): 1.35


Other Product Names / Synonyms: 2-Bromo-2-methylmalonic Acid Diethyl Ester

Formula: C₈H₁₃BrO₄
MW: 253.09 g/mol
Boiling Pt: 59 °C
Density: 1.35 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00009125
CAS Number: 29263-94-3
https://us.vwr.com/stibo/web/std.lang.all/64/11/28986411.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR