Dichlorophenylborane ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Phenylboron dichloride

D4367-5G D4367-25G
TCD4367-25GEA 432.73 USD
TCD4367-25G TCD4367-5G
Dichlorophenylborane ≥98.0% (by titrimetric analysis)
Dichlorophenylborane

CAS Number: 873-51-8
MDL Number: MFCD00013612
Molecular Formula: C6H5BCl2
Molecular Weight: 158.82
Purity/Analysis Method: 98.0% (T)
Form: Clear Liquid
Boiling point (°C): 177
Melting point (°C): 7
Flash Point (°C): 14
Specific Gravity (20/20): 1.20


Other Product Names / Synonyms: Phenylboron Dichloride

Formula: C₆H₅BCl₂
MW: 158.82 g/mol
Boiling Pt: 177 °C
Melting Pt: 7 °C
Density: 1.20 (20 °C)
Flash Pt: 14 °C
MDL Number: MFCD00013612
CAS Number: 873-51-8
UN: 2924
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/64/07/28986407.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR