(Difluoromethyl)trimethylsilane ≥97.0% (by GC)

Supplier: TCI America

D4364-5ML D4364-25ML D4364-1ML
TCD4364-1MLEA 54.79 USD
TCD4364-1ML TCD4364-5ML TCD4364-25ML
(Difluoromethyl)trimethylsilane ≥97.0% (by GC)
(Difluoromethyl)trimethylsilane

CAS Number: 65864-64-4
MDL Number: MFCD19381658
Molecular Formula: C4H10F2Si
Molecular Weight: 124.21
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 52
Specific Gravity (20/20): 0.91
Storage Temperature: 0-10°C

Formula: C₄H₁₀F₂Si
Boiling Pt: 52 °C
Density: 0.91 (20 °C)
MDL Number: MFCD19381658
CAS Number: 65864-64-4
https://us.vwr.com/stibo/web/std.lang.all/64/05/28986405.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR