2,2-Dimethyl-4-pentenoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2,2-Dimethylpent-4-enoic acid
D4354-1G
D4354-5G
TCD4354-1GEA
56.84
USD
TCD4354-1G
TCD4354-5G
2,2-Dimethyl-4-pentenoic acid ≥98.0% (by GC, titration analysis)
2,2-Dimethyl-4-pentenoic acid
CAS Number: 16386-93-9
MDL Number: MFCD00012304
Molecular Formula: C7H12O2
Molecular Weight: 128.17
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 108
Flash Point (°C): 88
Specific Gravity (20/20): 0.94
Formula:
C₇H₁₂O₂ MW: 128.17 g/mol Boiling Pt: 108 °C Density: 0.94 (20 °C) Flash Pt: 88 °C |
MDL Number:
MFCD00012304 CAS Number: 16386-93-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...