N-(9-Deoxy-epi-cinchonin-9-yl)picolinamide ≥94.0% (by HPLC)

Synonyms: N-[(9R)-Cinchonan-9-yl]picolinamide

D4305-100MG
TCD4305-100MGEA 250.86 USD
TCD4305-100MG
N-(9-Deoxy-epi-cinchonin-9-yl)picolinamide ≥94.0% (by HPLC)
N-(9-Deoxy-epi-cinchonin-9-yl)picolinamide

CAS Number: 1414851-55-0
Molecular Formula: C25H26N4O
Molecular Weight: 398.51
Purity/Analysis Method: 94.0% (HPLC)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Asymmetric Organocatalysts
Other Product Names / Synonyms: N-[(9R)-Cinchonan-9-yl]picolinamide

Formula: C₂₅H₂₆N₄O
MW: 398.51 g/mol
CAS Number: 1414851-55-0
https://us.vwr.com/stibo/web/std.lang.all/63/40/28986340.jpg

Order Now

Specification Test Results

Purity/Analysis Method 94.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR