1,2-Dimethoxypropane ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Propylene Glycol Dimethyl Ether

D4281-25ML
TCD4281-25MLEA 43 USD
TCD4281-25ML
1,2-Dimethoxypropane ≥98.0% (by GC)
1,2-Dimethoxypropane

CAS Number: 7778-85-0
MDL Number: MFCD00010331
Molecular Formula: C5H12O2
Molecular Weight: 104.15
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 96
Flash Point (°C): 1
Specific Gravity (20/20): 0.85


Other Product Names / Synonyms: Propylene Glycol Dimethyl Ether

Formula: C₅H₁₂O₂
MW: 104.15 g/mol
Boiling Pt: 96 °C
Density: 0.85 (20 °C)
Flash Pt: 1 °C
MDL Number: MFCD00010331
CAS Number: 7778-85-0
https://us.vwr.com/stibo/web/std.lang.all/62/63/28986263.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR