(2,5-Dimethylphenyl)acetonitrile ≥98.0% (by GC)
Supplier: TCI America
D4271-25G
D4271-5G
TCD4271-25GEA
158
USD
TCD4271-25G
TCD4271-5G
(2,5-Dimethylphenyl)acetonitrile ≥98.0% (by GC)
(2,5-Dimethylphenyl)acetonitrile
CAS Number: 16213-85-7
MDL Number: MFCD00013816
Molecular Formula: C10H11N
Molecular Weight: 145.21
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 79
Melting point (°C): 30
Other Product Names / Synonyms: 2,5-Dimethylbenzyl Cyanide
Formula:
C₁₀H₁₁N Boiling Pt: 79 °C Melting Pt: 30 °C |
MDL Number:
MFCD00013816 CAS Number: 16213-85-7 UN: 3439 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...