(2,4-Difluorophenyl)-4-piperidylmethanone oxime hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: (2,4-Difluorophenyl)(piperidin-4-yl)methanone oxime hydrochloride

D4269-5G
TCD4269-5GEA 70 USD
TCD4269-5G
(2,4-Difluorophenyl)-4-piperidylmethanone oxime hydrochloride ≥98.0% (by HPLC, titration analysis)
(2,4-Difluorophenyl)-4-piperidylmethanone oxime hydrochloride

CAS Number: 135634-18-3
MDL Number: MFCD11869788
Molecular Formula: C12H14F2N2O
Molecular Weight: 276.71
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal

Formula: C₁₂H₁₄F₂N₂O
MW: 276.71 g/mol
MDL Number: MFCD11869788
CAS Number: 135634-18-3
https://us.vwr.com/stibo/web/std.lang.all/62/35/28986235.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR