DAPT ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
(3,5-Difluorophenylacetyl)-Ala-Phg-OtBu
CAS Number: 208255-80-5
MDL Number: MFCD04974585
Molecular Formula: C23H26F2N2O4
Molecular Weight: 432.47
Purity/Analysis Method:
Form: Crystal
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Fluorinated Bioactive Compounds, Bioscience, Regenerative Medicine Research Reagents, Oligopeptides
Other Product Names / Synonyms: (3,5-Difluorophenylacetyl)-Ala-Phg-OtBu, (3,5-Difluorophenylacetyl)-L-alanyl-L-2-phenylglycine tert-Butyl Ester
Formula:
C₂₃H₂₆F₂N₂O₄ MW: 432.47 g/mol |
MDL Number:
MFCD04974585 CAS Number: 208255-80-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...