Diethyl [[2-(6-amino-9H-purin-9-yl)ethoxy]methyl]phosphonate ≥95.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 9-[2-(Diethoxyphosphorylmethoxy)ethyl]adenine[[2-(6-Amino-9H-purin-9-yl)ethoxy]methyl]phosphonic Acid Diethyl Ester

D4256-1G D4256-5G
TCD4256-1GEA 29.44 USD
TCD4256-1G TCD4256-5G
Diethyl [[2-(6-amino-9H-purin-9-yl)ethoxy]methyl]phosphonate ≥95.0% (by HPLC, titration analysis)
Diethyl [[2-(6-amino-9H-purin-9-yl)ethoxy]methyl]phosphonate

CAS Number: 116384-53-3
MDL Number: MFCD07783825
Molecular Formula: C12H20N5O4P
Molecular Weight: 329.30
Purity/Analysis Method: 95.0% (HPLC,T)
Form: Crystal
Melting point (°C): 136


Product Categories: Bioscience, Pharmacologically-Active Nucleosides and Nucleobases for Research and Experimental Use, Nucleosides and Their Analogs
Other Product Names / Synonyms: 9-[2-(Diethoxyphosphorylmethoxy)ethyl]adenine, [[2-(6-Amino-9H-purin-9-yl)ethoxy]methyl]phosphonic Acid Diethyl Ester

Formula: C₁₂H₂₀N₅O₄P
MW: 329.30 g/mol
Melting Pt: 136 °C
MDL Number: MFCD07783825
CAS Number: 116384-53-3
https://us.vwr.com/stibo/web/std.lang.all/62/20/28986220.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR