N-Octyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (Sulfobetaine-8, SB-8) ≥98.0% (by HPLC, total nitrogen) for biochemical research

Supplier: TCI America

Synonyms: 3-(N,N-Dimethyloctylammonio)propanesulfonate inner salt3-(N,N-Dimethyloctylammonio)propanesulphonate inner salt

D4247-5G
TCD4247-5GEA 161.39 USD
TCD4247-5G
N-Octyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (Sulfobetaine-8, SB-8) ≥98.0% (by HPLC, total nitrogen) for biochemical research
N-Octyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (Sulfobetaine-8, SB-8)

[for Biochemical Research]
CAS Number: 15178-76-4
MDL Number: MFCD00036912
Molecular Formula: C13H29NO3S
Molecular Weight: 279.44
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 205


Product Categories: Bioscience, Amphoteric Surfactants (for Biochemistry)
Other Product Names / Synonyms: 3-[Dimethyl(n-octyl)ammonio]propane-1-sulfonate, n-Octyl Sulfobetaine

Formula: C₁₃H₂₉NO₃S
Melting Pt: 205 °C
MDL Number: MFCD00036912
CAS Number: 15178-76-4
https://us.vwr.com/stibo/web/std.lang.all/62/12/28986212.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR