Daptomycin ≥94%

Supplier: TCI America

D4229-100MG
TCD4229-100MGEA 318.81 USD
TCD4229-100MG
Daptomycin ≥94%
Daptomycin

Daptomycin antiibiotic used to study bactericidal activity and membrane depolarization in Staphylococcus aureus.


  • Pale yellow crystals

Formula: C₇₂H₁₀₁N₁₇O₂₆
Storage Temperature: Freezer
MDL Number: MFCD08282794
CAS Number: 103060-53-3
https://us.vwr.com/stibo/web/std.lang.all/61/98/28986198.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR