Dexrazoxane ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: (S)-4,4'-(Propane-1,2-diyl)bis(piperazine-2,6-dione)(S)-1,2-Bis(3,5-dioxo-1-piperazinyl)propane

D4227-100MG
TCD4227-100MGEA 67.99 USD
TCD4227-100MG
Dexrazoxane ≥98.0% (by titrimetric analysis)
Dexrazoxane

CAS Number: 24584-09-6
MDL Number: MFCD00866449
Molecular Formula: C11H16N4O4
Molecular Weight: 268.27
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 195
Specific rotation [a]20/D: 11 deg (C=5, DMF)


Product Categories: Bioscience, Other Pharmaceutical Ingredients, Analytical Science, Complexones (Chelating Agents)
Other Product Names / Synonyms: (S)-4,4'-(Propane-1,2-diyl)bis(piperazine-2,6-dione), (S)-1,2-Bis(3,5-dioxo-1-piperazinyl)propane

Formula: C₁₁H₁₆N₄O₄
MW: 268.27 g/mol
Melting Pt: 195 °C
MDL Number: MFCD00866449
CAS Number: 24584-09-6
https://us.vwr.com/stibo/web/std.lang.all/61/95/28986195.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR