Duloxetine hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: (S)-(+)-N-Methyl-3-(1-naphthyloxy)-3-(2-thienyl)propylamine Hydrochloride

D4223-5G D4223-1G
TCD4223-5GEA 313 USD
TCD4223-5G TCD4223-1G
Duloxetine hydrochloride ≥98.0% (by HPLC, titration analysis)
Duloxetine hydrochloride

CAS Number: 136434-34-9
MDL Number: MFCD06407958
Molecular Formula: C18H19NOS
Molecular Weight: 333.87
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 164
Specific rotation [a]20/D: 122 deg (C=1, MeOH)


Product Categories: Bioscience, Neurotransmitter Agent Ingredients
Other Product Names / Synonyms: (S)-(+)-N-Methyl-3-(1-naphthyloxy)-3-(2-thienyl)propylamine Hydrochloride

Formula: C₁₈H₁₉NOS
MW: 333.87 g/mol
Melting Pt: 164 °C
MDL Number: MFCD06407958
CAS Number: 136434-34-9
https://us.vwr.com/stibo/web/std.lang.all/61/92/28986192.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR