1,5-Dichloro-2,4-dinitrobenzene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1,3,-Dichloro-4,6-dinitrobenzene

D4205-5G
TCD4205-5GEA 50.75 USD
TCD4205-5G
1,5-Dichloro-2,4-dinitrobenzene ≥98.0% (by GC)
1,5-Dichloro-2,4-dinitrobenzene

CAS Number: 3698-83-7
MDL Number: MFCD00024186
Molecular Formula: C6H2Cl2N2O4
Molecular Weight: 236.99
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 102

Formula: C₆H₂Cl₂N₂O₄
MW: 237 g/mol
Melting Pt: 102 °C
MDL Number: MFCD00024186
CAS Number: 3698-83-7
UN: 2811
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/61/75/28986175.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR