Dimethocaine ≥98.0% (by GC, titration analysis)
Synonyms:
Larocaine
CAS Number: 94-15-5
MDL Number: MFCD01697030
Molecular Formula: C16H26N2O2
Molecular Weight: 278.40
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 55
Product Categories: Bioscience, Other Pharmaceutical Ingredients
Other Product Names / Synonyms: Larocaine, 3-Diethylamino-2,2-dimethylpropyl 4-Aminobenzoate, 4-Aminobenzoic Acid 3-Diethylamino-2,2-dimethylpropyl Ester
Formula:
C₁₆H₂₆N₂O₂ MW: 278.40 g/mol Melting Pt: 55 °C |
MDL Number:
MFCD01697030 CAS Number: 94-15-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...