Desogestrel ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 13-Ethyl-11-methylene-18,19-dinor-17alpha-pregn-4-en-20-yn-17-ol

D4163-100MG
TCD4163-100MGEA 161.39 USD
TCD4163-100MG
Desogestrel ≥98.0% (by HPLC)
Desogestrel

CAS Number: 54024-22-5
MDL Number: MFCD00869346
Molecular Formula: C22H30O
Molecular Weight: 310.48
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 112
Specific rotation [a]20/D: 55 deg (C=1, CHCl3)


Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry), Bioscience, Other Pharmaceutical Ingredients, Other Steroids
Other Product Names / Synonyms: 13-Ethyl-11-methylene-18,19-dinor-17alpha-pregn-4-en-20-yn-17-ol

Formula: C₂₂H₃₀O
MW: 310.48 g/mol
Melting Pt: 112 °C
MDL Number: MFCD00869346
CAS Number: 54024-22-5
https://us.vwr.com/stibo/web/std.lang.all/61/01/28986101.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR