Diethyl-2,2'-thiodiglycolate ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 2,2'-Thiodiglycolic Acid Diethyl EsterDiethyl 2,2'-Thiodiacetate2,2'-Thiodiacetic Acid Diethyl Ester

D4147-5G
TCD4147-5GEA 39.86 USD
TCD4147-5G
Diethyl-2,2'-thiodiglycolate ≥97.0% (by GC)
Diethyl-2,2'-thiodiglycolate

CAS Number: 925-47-3
MDL Number: MFCD00087932
Molecular Formula: C8H14O4S
Molecular Weight: 206.26
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 105
Specific Gravity (20/20): 1.14


Product Categories: Synthetic Organic Chemistry, Sulfides
Other Product Names / Synonyms: 2,2'-Thiodiglycolic Acid Diethyl Ester, Diethyl 2,2'-Thiodiacetate, 2,2'-Thiodiacetic Acid Diethyl Ester

Formula: C₈H₁₄O₄S
MW: 206.26 g/mol
Boiling Pt: 105 °C
Density: 1.14 (20 °C)
MDL Number: MFCD00087932
CAS Number: 925-47-3
https://us.vwr.com/stibo/web/std.lang.all/60/85/28986085.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR