2,4-Dibromothiophene ≥90.0% (by GC)

Supplier: TCI America

D4146-1G D4146-5G
TCD4146-5GEA 97.44 USD
TCD4146-5G TCD4146-1G
2,4-Dibromothiophene ≥90.0% (by GC)
2,4-Dibromothiophene

CAS Number: 3140-92-9
MDL Number: MFCD00043889
Molecular Formula: C4H2Br2S
Molecular Weight: 241.93
Purity/Analysis Method: 90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 97
Flash Point (°C): 110
Specific Gravity (20/20): 2.17
Storage Temperature: 0-10°C

Formula: C₄H₂Br₂S
MW: 241.93 g/mol
Boiling Pt: 97 °C
Density: 2.17 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00043889
CAS Number: 3140-92-9
UN: 2810
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/60/84/28986084.jpg

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR