4,4'-Difluorobenzhydryl chloride ≥97.0% (by GC)
Supplier: TCI America
Synonyms:
Chloro bis-(4-fluorophenyl)methane
, 4,4-Difluorodiphenylmethylchloride
CAS Number: 27064-94-4
MDL Number: MFCD00044329
Molecular Formula: C13H9ClF2
Molecular Weight: 238.66
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 112
Flash Point (°C): 113
Specific Gravity (20/20): 1.28
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: Chlorobis(4-fluorophenyl)methane, alpha-Chloro-4,4'-difluorodiphenylmethane
Formula:
C₁₃H₉ClF₂ MW: 238.66 g/mol Boiling Pt: 112 °C Density: 1.28 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00044329 CAS Number: 27064-94-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...