1-(2,4-Xylyl)piperazin ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 1-(2,4-Dimethylphenyl)piperazine1-(2,4-Xylyl)piperazine

D4136-5G
TCD4136-5GEA 75 USD
TCD4136-5G
1-(2,4-Xylyl)piperazin ≥98.0% (by GC, titration analysis)
1-(2,4-Xylyl)piperazin

CAS Number: 1013-76-9
MDL Number: MFCD00023127
Molecular Formula: C12H18N2
Molecular Weight: 190.29
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 153
Flash Point (°C): 110
Specific Gravity (20/20): 1.01


Other Product Names / Synonyms: 1-(2,4-Xylyl)piperazine

Formula: C₁₂H₁₈N₂
MW: 190.29 g/mol
Boiling Pt: 153 °C
Density: 1.01 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00023127
CAS Number: 1013-76-9
https://us.vwr.com/stibo/web/std.lang.all/60/74/28986074.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR