9,10-Di(2-naphthyl)anthracene ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
9,10-Di(naphthalen-2-yl)anthracene
D4127-1G
TCD4127-1GEA
176.61
USD
TCD4127-1G
9,10-Di(2-naphthyl)anthracene ≥98.0% (by HPLC)
9,10-Di(2-naphthyl)anthracene
CAS Number: 122648-99-1
MDL Number: MFCD00028944
Molecular Formula: C34H22
Molecular Weight: 430.55
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 357
Formula:
C₃₄H₂₂ MW: 430.55 g/mol Melting Pt: 357 °C |
MDL Number:
MFCD00028944 CAS Number: 122648-99-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...