Doxazosin mesylate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 1-(4-Amino-6,7-dimethoxy-2-quinazolinyl)-4-[(1,4-benzodioxan-2-yl)carbonyl]piperazine Methanesulfonate

D4126-1G D4126-100MG
TCD4126-1GEA 278.11 USD
TCD4126-1G TCD4126-100MG
Doxazosin mesylate ≥98.0% (by HPLC, total nitrogen)
Doxazosin mesylate

CAS Number: 77883-43-3
MDL Number: MFCD00216023
Molecular Formula: C23H25N5O5
Molecular Weight: 547.58
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 269


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Antihypertensive Ingredients
Other Product Names / Synonyms: 1-(4-Amino-6,7-dimethoxy-2-quinazolinyl)-4-[(1,4-benzodioxan-2-yl)carbonyl]piperazine Methanesulfonate

Formula: C₂₃H₂₅N₅O₅
MW: 547.58 g/mol
Melting Pt: 269 °C
MDL Number: MFCD00216023
CAS Number: 77883-43-3
https://us.vwr.com/stibo/web/std.lang.all/60/64/28986064.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR