Dimethyldioctadecylammonium Iodide ≥98.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: Dimethyldistearylammonium Iodide

D4124-5G
TCD4124-5GEA 171.46 USD
TCD4124-5G
Dimethyldioctadecylammonium Iodide ≥98.0% (by total nitrogen basis)
Dimethyldioctadecylammonium Iodide

CAS Number: 7206-39-5
Molecular Formula: C38H80IN
Molecular Weight: 677.97
Purity/Analysis Method: 98.0% (N)
Form: Crystal
Melting point (°C): 154


Other Product Names / Synonyms: Dimethyldistearylammonium Iodide

Formula: C₃₈H₈₀IN
MW: 677.97 g/mol
Melting Pt: 154 °C
CAS Number: 7206-39-5
https://us.vwr.com/stibo/web/std.lang.all/60/62/28986062.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR