3,6-Dichloropicolinic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3,6-Dichloropyridine-2-carboxylic acid

D4117-1G
TCD4117-1GEA 42.43 USD
TCD4117-1G
3,6-Dichloropicolinic acid ≥98.0% (by GC, titration analysis)
3,6-Dichloropicolinic acid

CAS Number: 1702-17-6
MDL Number: MFCD00078655
Molecular Formula: C6H3Cl2NO2
Molecular Weight: 192.00
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 151


Other Product Names / Synonyms: Clopyralid, 3,6-Dichloropicolinic Acid

Formula: C₆H₃Cl₂NO₂
MW: 192.00
Melting Pt: 151 °C
MDL Number: MFCD00078655
CAS Number: 1702-17-6
https://us.vwr.com/stibo/web/std.lang.all/60/54/28986054.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR