Diethyl-3,3'-iminodipropionate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Diethyl 3,3'-azanediyldipropanoate3,3'-Iminodipropionic Acid Diethyl Ester

D4111-1G D4111-5G
TCD4111-5GEA 374.98 USD
TCD4111-5G TCD4111-1G
Diethyl-3,3'-iminodipropionate ≥98.0% (by GC, titration analysis)
Diethyl-3,3'-iminodipropionate

CAS Number: 3518-88-5
MDL Number: MFCD14705114
Molecular Formula: C10H19NO4
Molecular Weight: 217.27
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 139
Specific Gravity (20/20): 1.04
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 3,3'-Iminodipropionic Acid Diethyl Ester

Formula: C₁₀H₁₉NO₄
MW: 217.27 g/mol
Boiling Pt: 139 °C
Density: 1.04 (20 °C)
MDL Number: MFCD14705114
CAS Number: 3518-88-5
https://us.vwr.com/stibo/web/std.lang.all/60/49/28986049.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR