3-(Dimethylamino)acrolein ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-(Dimethylamino)acrylaldehyde3-(Dimethylamino)propenal3-(Dimethylamino)-2-propenal

D4092-5G D4092-25G
TCD4092-25GEA 199.4 USD
TCD4092-25G TCD4092-5G
3-(Dimethylamino)acrolein ≥98.0% (by GC, titration analysis)
3-(Dimethylamino)acrolein

CAS Number: 927-63-9
MDL Number: MFCD00006999
Molecular Formula: C5H9NO
Molecular Weight: 99.13
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 91
Flash Point (°C): 113
Specific Gravity (20/20): 0.99


Other Product Names / Synonyms: 3-(Dimethylamino)acrylaldehyde, 3-(Dimethylamino)-2-propenal

Formula: C₅H₉NO
MW: 99.13 g/mol
Boiling Pt: 91 °C
Density: 0.99 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00006999
CAS Number: 927-63-9
https://us.vwr.com/stibo/web/std.lang.all/60/36/28986036.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR