1,3-Di-9-carbazolylbenzene ≥98.0% (by HPLC, total nitrogen), purified by sublimation
Supplier: TCI America
Synonyms:
1,3-Bis(carbazol-9-yl)benzene
D4087-1G
TCD4087-1GEA
220
USD
TCD4087-1G
1,3-Di-9-carbazolylbenzene ≥98.0% (by HPLC, total nitrogen), purified by sublimation
1,3-Di-9-carbazolylbenzene
CAS Number: 550378-78-4
MDL Number: MFCD09836836
Molecular Formula: C30H20N2
Molecular Weight: 408.50
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 184
Product Categories: Materials Chemistry, Organic Light-Emitting Diode (OLED) Materials, Highly Purified Reagents
Other Product Names / Synonyms: 1,3-Bis(carbazol-9-yl)benzene
Formula:
C₃₀H₂₀N₂ MW: 408.50 g/mol Melting Pt: 184 °C |
MDL Number:
MFCD09836836 CAS Number: 550378-78-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...