(R)-4-(2-Hydroxyethyl)-2,2-diisopropyl-1,3-dioxolane ≥95.0%
Supplier: TCI America
Synonyms:
(R)-2-(2,2-Diisopropyl-1,3-dioxolan-4-yl)ethanol
D4076-1G
TCD4076-1GEA
247.55
USD
TCD4076-1G
(R)-4-(2-Hydroxyethyl)-2,2-diisopropyl-1,3-dioxolane ≥95.0%
(R)-4-(2-Hydroxyethyl)-2,2-diisopropyl-1,3-dioxolane
Molecular Weight: 202.29
Purity/Analysis Method:
Form: Clear Liquid
Specific rotation [a]20/D: -5 deg (C=15, CHCl3)
Molecular Formula: C11H22O3
Other Product Names / Synonyms: (R)-2-(2,2-Diisopropyl-1,3-dioxolan-4-yl)ethanol
Formula:
C₁₁H₂₂O₃ MW: 202.29 g/mol |
CAS Number:
1820574-15-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...