Dimethyl-trans-2,3-dibromobutenedioate ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
trans-2,3-Dibromobutenedioic Acid Dimethyl Ester
, 2,3-Dibromofumaric Acid Dimethyl Ester
, Dimethyl 2,3-Dibromofumarate
D4073-5G
TCD4073-5GEA
356.11
USD
TCD4073-5G
Dimethyl-trans-2,3-dibromobutenedioate ≥98.0% (by GC)
Dimethyl-trans-2,3-dibromobutenedioate
CAS Number: 116631-94-8
Molecular Formula: C6H6Br2O4
Molecular Weight: 301.92
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 90
Melting point (°C): 40
Storage Temperature: 0-10°C
Other Product Names / Synonyms: trans-2,3-Dibromobutenedioic Acid Dimethyl Ester, Dimethyl 2,3-Dibromofumarate, 2,3-Dibromofumaric Acid Dimethyl Ester
Formula:
C₆H₆Br₂O₄ MW: 301.92 g/mol Boiling Pt: 90 °C Melting Pt: 40 °C |
CAS Number:
116631-94-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...