2,5-Difluorobenzenesulfonamide ≥98.0% (by GC, total nitrogen)
Supplier: TCI America
D4060-1G
D4060-5G
TCD4060-1GEA
82
USD
TCD4060-1G
TCD4060-5G
2,5-Difluorobenzenesulfonamide ≥98.0% (by GC, total nitrogen)
2,5-Difluorobenzenesulphonamide
CAS Number: 120022-63-1
MDL Number: MFCD00798115
Molecular Formula: C6H5F2NO2S
Molecular Weight: 193.17
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 137
Formula:
C₆H₅F₂NO₂S MW: 193.17 g/mol Melting Pt: 137 °C |
MDL Number:
MFCD00798115 CAS Number: 120022-63-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...