4-(Dimethylamino)thiophenol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D4047-1G
TCD4047-1GEA 144.97 USD
TCD4047-1G
4-(Dimethylamino)thiophenol ≥98.0% (by GC, titration analysis)
4-(Dimethylamino)thiophenol

CAS Number: 4946-22-9
MDL Number: MFCD01658123
Molecular Formula: C8H11NS
Molecular Weight: 153.24
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Boiling point (°C): 260
Melting point (°C): 33


Product Categories: Materials Chemistry, Self-Assembled Monolayer (SAM) Forming Agents
Other Product Names / Synonyms: 4-(Dimethylamino)thiophenol, 4-Mercapto-N,N-dimethylaniline

Formula: C₈H₁₁NS
Boiling Pt: 260 °C
Melting Pt: 33 °C
MDL Number: MFCD01658123
CAS Number: 4946-22-9
https://us.vwr.com/stibo/web/std.lang.all/59/94/28985994.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR