1,3-Dichloro-5-fluorobenzene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 3,5-Dichlorofluorobenzene

D4046-5G D4046-25G
TCD4046-5GEA 72.54 USD
TCD4046-5G TCD4046-25G
1,3-Dichloro-5-fluorobenzene ≥98.0% (by GC)
1,3-Dichloro-5-fluorobenzene

CAS Number: 1435-46-7
MDL Number: MFCD00060652
Molecular Formula: C6H3Cl2F
Molecular Weight: 164.99
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 162
Specific Gravity (20/20): 1.39

Formula: C₆H₃Cl₂F
MW: 164.9900 g/mol
Boiling Pt: 162 °C
Density: 1.39 (20 °C)
MDL Number: MFCD00060652
CAS Number: 1435-46-7
https://us.vwr.com/stibo/web/std.lang.all/59/93/28985993.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR