2,5-Difluorotoluene ≥98.0% (by GC)

Synonyms: 1,4-Difluoro-2-methylbenzene

D4042-25G D4042-5G
TCD4042-5GEA 44.88 USD
TCD4042-5G TCD4042-25G
2,5-Difluorotoluene ≥98.0% (by GC)
2,5-Difluorotoluene

CAS Number: 452-67-5
MDL Number: MFCD00000347
Molecular Formula: C7H6F2
Molecular Weight: 128.12
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 117
Melting point (°C): -35
Flash Point (°C): 12
Specific Gravity (20/20): 1.13

Formula: C₇H₆F₂
MW: 128.12 g/mol
Boiling Pt: 117 °C
Melting Pt: –35 °C
Density: 1.13 (20 °C)
Flash Pt: 12 °C
MDL Number: MFCD00000347
CAS Number: 452-67-5
UN: 2388
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/59/91/28985991.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR