2,5-Dibromothieno[2,3-b]thiophene ≥98.0% (by GC)
Supplier: TCI America
D4032-1G
TCD4032-1GEA
491.05
USD
TCD4032-1G
2,5-Dibromothieno[2,3-b]thiophene ≥98.0% (by GC)
2,5-Dibromothieno[2,3-b]thiophene
CAS Number: 25121-86-2
Molecular Formula: C6H2Br2S2
Molecular Weight: 298.01
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 96
Storage Temperature:
Formula:
C₆H₂Br₂S₂ MW: 298.01 g/mol Melting Pt: 96 °C |
CAS Number:
25121-86-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...