2,5-Dibromoselenophene ≥98.0% (by GC)

Supplier: TCI America

D4028-1G D4028-5G
TCD4028-5GEA 445 USD
TCD4028-5G TCD4028-1G
2,5-Dibromoselenophene ≥98.0% (by GC)
2,5-Dibromoselenophene

CAS Number: 1755-36-8
Molecular Formula: C4H2Br2Se
Molecular Weight: 288.83
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 104
Specific Gravity (20/20): 2.50
Storage Temperature: 0-10°C

Formula: C₄H₂Br₂Se
MW: 288.83 g/mol
Boiling Pt: 104 °C
Density: 2.50 (20 °C)
CAS Number: 1755-36-8
https://us.vwr.com/stibo/web/std.lang.all/59/73/28985973.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR