Dantrolene sodium salt hydrate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 1-[5-(4-Nitrophenyl)furfurylideneamino]hydantoin Sodium Salt

D3996-5G D3996-1G
TCD3996-5GEA 387.73 USD
TCD3996-5G TCD3996-1G
Dantrolene sodium salt hydrate ≥98.0% (by HPLC, titration analysis)
Dantrolene sodium salt hydrate

CAS Number: 14663-23-1
MDL Number: MFCD00079130
Molecular Formula: C14H10N4O5
Molecular Weight: 336.24
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal


Product Categories: Bioscience, Other Pharmaceutical Ingredients, Ryanodine Receptor (RyR) Antagonists
Other Product Names / Synonyms: 1-[5-(4-Nitrophenyl)furfurylideneamino]hydantoin Sodium Salt

Formula: C₁₄H₁₀N₄O₅
MW: 336.24 g/mol
MDL Number: MFCD00079130
CAS Number: 14663-23-1
https://us.vwr.com/stibo/web/std.lang.all/59/40/28985940.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR