Dimethyl-1,3-benzodithiol-2-ylphosphonate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1,3-Benzodithiol-2-ylphosphonic Acid Dimethyl Ester2-Dimethoxyphosphinyl-1,3-benzodithiole

D3992-1G
TCD3992-1GEA 200.88 USD
TCD3992-1G
Dimethyl-1,3-benzodithiol-2-ylphosphonate ≥98.0% (by GC)
Dimethyl-1,3-benzodithiol-2-ylphosphonate

CAS Number: 62217-35-0
Molecular Formula: C9H11O3PS2
Molecular Weight: 262.28
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 122
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Sulfides, Carbonyl Olefination, Materials Chemistry, Molecular Conductors
Other Product Names / Synonyms: 1,3-Benzodithiol-2-ylphosphonic Acid Dimethyl Ester, 2-Dimethoxyphosphinyl-1,3-benzodithiole

Formula: C₉H₁₁O₃PS₂
MW: 262.28 g/mol
Melting Pt: 122 °C
CAS Number: 62217-35-0
https://us.vwr.com/stibo/web/std.lang.all/59/36/28985936.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR