2,2-Dimethylbutyryl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2,2-Dimethylbutyrylchloride

D3987-250G D3987-25G
TCD3987-250GEA 293.55 USD
TCD3987-250G TCD3987-25G
2,2-Dimethylbutyryl chloride ≥98.0% (by GC, titration analysis)
2,2-Dimethylbutyryl chloride

CAS Number: 5856-77-9
MDL Number: MFCD03093071
Molecular Formula: C6H11ClO
Molecular Weight: 134.60
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 132
Flash Point (°C): 29
Specific Gravity (20/20): 0.98

Formula: C₆H₁₁ClO
MW: 134.61 g/mol
Boiling Pt: 132 °C
Density: 0.98 (20 °C)
Flash Pt: 29 °C
MDL Number: MFCD03093071
CAS Number: 5856-77-9
https://us.vwr.com/stibo/web/std.lang.all/59/33/28985933.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR