2,6-Dichloro-5-fluoronicotinonitrile ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2,6-Dichloro-5-fluoro-3-pyridinecarbonitrile2,6-Dichloro-3-cyano-5-fluoropyridine

D3972-25G D3972-5G
TCD3972-5GEA 18.27 USD
TCD3972-5G TCD3972-25G
2,6-Dichloro-5-fluoronicotinonitrile ≥98.0% (by GC)
2,6-Dichloro-5-fluoronicotinonitrile

CAS Number: 82671-02-1
MDL Number: MFCD01863289
Molecular Formula: C6HCl2FN2
Molecular Weight: 190.99
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 91


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2,6-Dichloro-3-cyano-5-fluoropyridine

Formula: C₆HCl₂FN₂
MW: 190.99 g/mol
Melting Pt: 91 °C
MDL Number: MFCD01863289
CAS Number: 82671-02-1
UN: 3439
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/59/16/28985916.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR