1,2-Dipalmitoyl-sn-glycero-3-phosphocholine ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
L-α-Lecithin, Dipalmitoyl
CAS Number: 63-89-8
MDL Number: MFCD00036903
Molecular Formula: C40H80NO8P
Molecular Weight: 734.05
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 220
Specific rotation [a]20/D: 7.0 deg (C=2, CHCl3)
Storage Temperature:
Product Categories: Bioscience, Lipids, Drug Delivery Systems (DDS) Research Reagents
Other Product Names / Synonyms: 1,2-Dihexadecanoyl-sn-glycero-3-phosphocholine, 2,3-Bis(palmitoyloxy)propyl [2-(Trimethylammonio)ethyl] Phosphate, L-DPPC
Formula:
C₄₀H₈₀NO₈P MW: 734.05 g/mol Melting Pt: 220 °C |
MDL Number:
MFCD00036903 CAS Number: 63-89-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...