N,N'-Diisopropylethylenediamine ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: N,N’-Diisopropylethylenediamine

D3923-25G D3923-5G
TCD3923-25GEA 160.37 USD
TCD3923-25G TCD3923-5G
N,N'-Diisopropylethylenediamine ≥97.0% (by GC, titration analysis)
N,N'-Diisopropylethylenediamine

CAS Number: 4013-94-9
MDL Number: MFCD00008864
Molecular Formula: C8H20N2
Molecular Weight: 144.26
Purity/Analysis Method: 97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 170
Flash Point (°C): 52
Specific Gravity (20/20): 0.80

https://us.vwr.com/stibo/web/std.lang.all/58/69/28985869.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR