Daidzin ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 7-(beta-D-Glucopyranosyloxy)-4'-hydroxyisoflavone

D3920-25MG
TCD3920-25MGEA 146.11 USD
TCD3920-25MG
Daidzin ≥98.0% (by HPLC)
Daidzin

CAS Number: 552-66-9
MDL Number: MFCD00017466
Molecular Formula: C21H20O9
Molecular Weight: 416.38
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 242
Specific rotation [a]20/D: -39 deg (C=0.5, DMSO)
Lambda max.: 297 nmStorage Temperature: 0-10°C


Product Categories: Bioscience, Glycosides, Glucose, Glycosides, Inhibitors (for Pharmacological Research), Crude Medicine Ingredients for Research and Experimental Use, Aromatic Polyketides, Polyphenols (Phytochemicals)
Other Product Names / Synonyms: 7-(beta-D-Glucopyranosyloxy)-4'-hydroxyisoflavone

Formula: C₂₁H₂₀O₉
MW: 416.38 g/mol
Melting Pt: 242 °C
MDL Number: MFCD00017466
CAS Number: 552-66-9
https://us.vwr.com/stibo/web/std.lang.all/58/67/28985867.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR