Disodium prednisolone-21-phosphate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Prednisolone 21-Phosphate Disodium Salt

D3918-5G
TCD3918-5GEA 301.46 USD
TCD3918-5G
Disodium prednisolone-21-phosphate ≥98.0% (by HPLC, titration analysis)
Disodium prednisolone-21-phosphate

CAS Number: 125-02-0
MDL Number: MFCD00083682
Molecular Formula: C21H29O8P
Molecular Weight: 484.39
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: 99 deg (C=1, pH 7.0 Buffer sol.)
Lambda max.: 243 nm (MeOH)


Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents, Bone Metabolism Research Reagents, Anti-inflammatory Ingredients, Anti-inflammatory Ingredients, Pregnanes (Glucocorticoids)
Other Product Names / Synonyms: Prednisolone 21-Phosphate Disodium Salt

Formula: C₂₁H₂₉O₈P
MW: 484.39 g/mol
MDL Number: MFCD00083682
CAS Number: 125-02-0
https://us.vwr.com/stibo/web/std.lang.all/58/65/28985865.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR