Nitrofen ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2,4-Dichloro-1-(4-nitrophenoxy)benzene

D3912-5G
TCD3912-5GEA 177 USD
TCD3912-5G
Nitrofen ≥98.0% (by GC)
Nitrofen

CAS Number: 1836-75-5
MDL Number: MFCD00128026
Molecular Formula: C12H7Cl2NO3
Molecular Weight: 284.09
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 180
Melting point (°C): 71
Flash Point (°C): 200


Other Product Names / Synonyms: Nitrofen

Formula: C₁₂H₇Cl₂NO₃
MW: 284.1 g/mol
Boiling Pt: 180 °C
Melting Pt: 71 °C
Flash Pt: 200 °C
MDL Number: MFCD00128026
CAS Number: 1836-75-5
https://us.vwr.com/stibo/web/std.lang.all/58/60/28985860.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR